Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ SAX Plates

Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE well plates and individual wells.

$671.00

Specifications

For Use With (Application) Extraction of Weak Acids
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep SAX
Type 96-well Plate
View More Specs

Products 5
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
03-251-206
View Documents Promotion Details
Thermo Scientific™
60300567
SAX 50 mg/1 mL 96 Removable Well Plate, 1 pk
Each for $671.00
Only null left
Add to Cart
 
60-307-242
View Documents Promotion Details
Thermo Scientific™
60307242
SAX 25 mg/2 mL 96 Fixed Well Plate, 1 pk
Pack for $671.00
Only null left
Add to Cart
 
60-307-243
View Documents Promotion Details
Thermo Scientific™
60307243
SAX 50 mg/2 mL 96 Fixed Well Plate, 1 pk
Pack for $671.00
Only null left
Add to Cart
 
03-251-204
View Documents
Thermo Scientific™
60300565
SAX 10 mg/1 mL 96 Removable Well Plate, 1 pk N/A N/A N/A
03-251-205
View Documents
Thermo Scientific™
60300566
SAX 25 mg/1 mL 96 Removable Well Plate, 1 pk N/A N/A N/A
Description

Description

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ SAX Plates feature a strong anion exchange sorbent for extraction of weak acids. Typical applications include:

  • Isolation of anionic proteins
  • Removal of acidic food pigments
  • Isolation of phenolic compounds
  • Nucleic acids, nucleotides and surfactants

HyperSep SAX Strong Anion Exchanger SPE 96-Well Plates and Individual Wells are ideally suited for the extraction of carboxylic and other weak acids. The chemistry extracts negatively charged compounds from both aqueous and nonaqueous solutions.
  • Amine group masks the effect of the carbon chain of the functional group
  • Selectivity can be tuned by choice of buffer

HyperSep SPE Cartridges and Plates

Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Specifications

Specifications

Extraction of Weak Acids
HyperSep
96-well Plate
40 to 60 μm
HyperSep SAX
1 mL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.