Learn More
Thermo Scientific™ HyperSep™ SPE Plates
Description
Innovative HyperSep™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ HyperSep™ Solid Phase Extraction Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, HyperSep SPE Well Plates deliver unparalleled performance, including:
- Higher levels of reproducibility
- Lower elution volumes
- High extract cleanliness
HyperSep Solid Phase Extraction Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You'll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
- 10 mg/2 mL 96 well plate format
- Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
- HyperSep products are also available in cartridge format.
Specifications
Specifications
| Bed Weight | 10 mg |
| For Use With (Application) | Bioanalytical Applications - Weak Ion-Exchange Retention of Strong Acidic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds |
| Product Line | HyperSep |
| Type | 96-well Plate |
| Volume (Metric) Well | 1 mL |
| Includes | 10 mg/1 mL 96-well plate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.