Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns

Catalog No. 30205254030 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
150 mm
200 mm
250 mm
Diameter (Metric):
4 mm
4.6 mm
Particle Size:
5 μm
10 μm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
30-205-254030 250 mm 4 mm 5 μm
30-205-054630 50 mm 4.6 mm 5 μm
03-050-127 150 mm 4.6 mm 5 μm
03-050-128 250 mm 4.6 mm 5 μm
03-150-545 10 mm 4 mm 5 μm
03-260-199 200 mm 4.6 mm 10 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 30-205-254030 Supplier Thermo Scientific™ Supplier No. 30205254030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use as a general-purpose C8 phase for acidic and neutral analytes.

For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.

Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable

Ordering Information:

Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4 mm
Length (Metric) 250 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 120 Å
Surface Area 170 m²/g
Temperature 60°C
USP Type L7
Stationary Phase C8
Product Line Hypersil Classical
Packing Material Spherical, Fully Porous Type A Silica
Carbon Load 6.5%
Column Type Reversed Phase
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.