Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hypromellose, Substitution Type 2910, 4000 mPa.s, USP, 7-12%, Spectrum™ Chemical

Click to view available options
Quantity:
125 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Hypromellose, Substitution Type 2910, 4000 mPa.s, USP is an artificial tear used to relieve dry eye conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Quantity | 2.5 kg |
Percent Purity | 7 to 12% |
Grade | USP |
Ignition Residue | 1.5% |
Loss on Drying | 5% |
Packaging | Poly Pail |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction