Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hypusine Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS1064I2MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1064I2MI 25 μL
ABS1064I1MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS1064I2MI Supplier MilliporeSigma Supplier No. ABS1064I25UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hypusine Polyclonal antibody specifically detects Hypusine in Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hypusine
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Alias N -4-amino-2-hydroxybutyl(lysine)
Host Species Rabbit
Immunogen Synthetic hypusine-containing peptide C-Ahx-STSKTG(hypusine)HGHAKV-amide.
Molecular Weight of Antigen ∼45 kDa observed (GFP-elf-5A). Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects hypusinated form of elF-5A.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.