Learn More
BUCHI I-100 Interface
SDP
The I-100 is an easy to use interface to conveniently regulate vacuum. As a central interface of the Rotavapor R-100, it increases the efficiency of the evaporation process.
Supplier:  BUCHI 11I100001
 View more versions of this product
Catalog No. 05-405-123
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Specifications
CE
Vacuum Controller
Power Pack, and Integrated Holder to Fasten to Lab-Stand
160 mm
PBT
Vacuum Pump Model V-100
I-100
24 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.