Learn More
BUCHI I-100 Interface
The I-100 is an easy to use interface to conveniently regulate vacuum. As a central interface of the Rotavapor R-100, it increases the efficiency of the evaporation process.
Specifications
Material | PBT |
---|---|
Type | Vacuum Controller |
For Use With (Equipment) | Vacuum Pump Model V-100 |
Model | I-100 |
Voltage | 24 V |
Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
05-405-192
![]() |
BUCHI
11I100003 |
Vacuum Pump Model V-100 | Power Pack, Valve Unit, and Holder to Fasten to Lab-Stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
05-405-123
![]() |
BUCHI
11I100001 |
Vacuum Pump Model V-100 | Power Pack, and Integrated Holder to Fasten to Lab-Stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
05-405-122
![]() |
BUCHI
11I100002 |
Vacuum Pump Model V-100 | Power Pack, Communication Cable (Mini-DIN), and Holder to Fasten to Lab-Stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Functional design fulfills essential needs of vacuum regulation
- Saves resources thanks to control of vacuum pump
- High longevity due to robust and durable design
- User-friendly and intuitive handling due to self-explaining user interface
- Easy and accurate vacuum regulation
- Relevant information is displayed at a glance
- High reliability due to centralized control of major process parameters
- Full process control thanks to parameter monitoring
- Proven technology for major applications
Specifications
PBT | |
Vacuum Pump Model V-100 | |
24 V |
Vacuum Controller | |
I-100 |