Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-A/I-E Mouse, anti-Mouse, R718, Clone: 2G9, BD OptiBuild™

Catalog No. BDB752163 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752163 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752163 Supplier BD Biosciences Supplier No. 752163
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2G9 monoclonal antibody reacts with the I-Ad and I-Ed MHC class II alloantigens. It also reacts with transfectants expressing I-Eb and I-Ek and with cells from mice of the H-2p and H-2q haplotypes. This antibody is non-reactive with cells from SJL (H-2s) and NOD (H-2g7) mice. Flow cytometric analysis indicates that the 2G9 and M5/114.15.2 monoclonal antibodies have comparable reactivity on cells from mice with I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek alloantigens.

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-A/I-E
Applications Flow Cytometry
Classification Monoclonal
Clone 2G9
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.