Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-A/I-E Rat anti-Mouse, BUV737, Clone: M5/114.15.2 (also known as M5/114), BD Horizon™

Catalog No. BDB569176 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569176 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569176 Supplier BD Biosciences Supplier No. 569176
Add to Cart
Add to Cart

Rat Monoclonal Antibody

I-A/I-E Monoclonal antibody specifically detects I-A/I-E in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen I-A/I-E
Applications Flow Cytometry
Classification Monoclonal
Clone M5/114.15.2 (also known as M5/114)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 737
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias H-2I; I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek MHC class II alloAgs; Ia Ag; M5/114; MHC II
Host Species Rat
Immunogen Activated C57BL/6 Mouse Spleen Cells
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111861
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.