Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-A/I-E Rat anti-Mouse, RY586, Clone: M5/114.15.2 (also known as M5/114), BD Horizon™
Rat Monoclonal Antibody
$140.60 - $352.00
Specifications
Antigen | I-A/I-E |
---|---|
Clone | M5/114.15.2 (also known as M5/114) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
I-A/I-E Monoclonal antibody specifically detects I-A/I-E in Mouse samples. It is validated for Flow cytometrySpecifications
I-A/I-E | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
111861 | |
IgG2b κ | |
Affinity Chromatography |
M5/114.15.2 (also known as M5/114) | |
Flow Cytometry | |
RY586 | |
Rat | |
Mouse | |
H-2I; I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek MHC class II alloAgs; Ia Ag; M5/114; MHC II | |
Activated C57BL/6 Mouse Spleen Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title