Learn More
Invitrogen™ I-Ab/I-Ad Monoclonal Antibody (25-9-17S), PE

Description
MA5-17777 targets I-Ab/I-Ad in FACS applications and shows reactivity with Mouse samples. The MA5-17777 immunogen is c3H.SW splenocytes.
Specifications
Specifications
| Antigen | I-Ab/I-Ad |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 25-9-17S |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Gene | H2-Ab1 |
| Gene Accession No. | P01921, P06342, P06343, P06344, P06345, P06346, P14483 |
| Gene Alias | Abeta; AI845868; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; Ia2; Ia-2; IAb; I-Abeta; major histocompatibility complex class II beta chain; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.