Learn More
Invitrogen™ I-Ab Monoclonal Antibody (28-5-16S)

Description
MA1-70065 detects I-Ab in mouse samples. MA1-70065 has been successfully used in flow cytometry procedures. The MA1-70065 immunogen is H2-I / Ab.
Specifications
Specifications
| Antigen | I-Ab |
| Applications | Cytotoxicity Assay, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 28-5-16S |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | H2-Ab1 |
| Gene Accession No. | P01921, P06342, P06343, P06344, P06345, P06346, P14483 |
| Gene Alias | Abeta; AI845868; CELIAC1; DADB-249P12.2; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2Eb; H-2Eb; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; HLA-DQB; Ia2; Ia-2; Ia4; Ia-4; IAb; I-Abeta; IDDM1; major histocompatibility complex class II beta chain; MGC163794; MGC163796; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.