Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-Ad Mouse, anti-Mouse, R718, Clone: 34-5-3, BD OptiBuild™

Catalog No. BDB751908 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751908 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751908 Supplier BD Biosciences Supplier No. 751908
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 34-5-3 antibody reacts with the β chain of the I-A[d] MHC class II alloantigen. It cross-reacts with I-A[b] and with cells from mice of the H-2[p] and H-2[q] haplotypes. Reactivity with other haplotypes (e.g., a, f, k, r, s) has not been observed. The strain distribution of the antigen recognized by this reagent is similar to that of anti-I-A[b] mAb 25-9-17 (Cat. Nos. 553603, 553604, and 553605).

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ad
Applications Flow Cytometry
Classification Monoclonal
Clone 34-5-3
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.