Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ I-Ak Monoclonal Antibody (14V.18), FITC
GREENER_CHOICE

Catalog No. PIMA517773
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517773 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517773 Supplier Invitrogen™ Supplier No. MA517773
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17773 targets I-Ak in FACS applications and shows reactivity with Mouse samples. The MA5-17773 immunogen is a.TL spleen.

MHC Class II (I-Ak) serves as a critical component of the immune system, facilitating the recognition and response to extracellular pathogens and foreign substances. Its primary role is to present peptides derived from extracellular proteins to T cells for immune recognition. MHC Class II (I-Ak) is composed of an alpha and beta chain, both of which contribute to the peptide-binding groove. The location of MHC Class II (I-Ak) is restricted to antigen-presenting cells, such as dendritic cells, macrophages, and B cells. These cells are responsible for capturing and processing antigens from the extracellular environment. MHC Class II (I-Ak) consists of an extracellular domain, a transmembrane domain, and a cytoplasmic tail. The extracellular domain is responsible for interacting with the antigenic peptides, while the transmembrane domain anchors the MHC Class II molecule to the cell membrane. The cytoplasmic tail plays a role in intracellular signaling events. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ak
Applications Flow Cytometry
Classification Monoclonal
Clone 14V.18
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene H2-Aa
Gene Accession No. P01910, P04227, P04228, P14434, P14435, P14436, P14437, P14438, P23150
Gene Alias A alpha; Aalpha; H-2 class II histocompatibility antigen, A-B alpha chain; H-2 class II histocompatibility antigen, A-F alpha chain; H-2 class II histocompatibility antigen, A-K alpha chain; H-2 class II histocompatibility antigen, A-Q alpha chain; H-2 class II histocompatibility antigen, A-R alpha chain; H-2 class II histocompatibility antigen, A-S alpha chain; H-2 class II histocompatibility antigen, A-U alpha chain; H2Aa; H-2Aa; H2-Aa; histocompatibility 2, class II antigen A, alpha; Ia1; Ia-1; IAalpha; I-Aalpha; major histocompatibility protein class II alpha chain; MHC class II antigen A alpha isoform-004; MHC-II I-A(d) alpha
Gene Symbols H2-Aa
Host Species Mouse
Immunogen A.TL spleen.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14960
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.