Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-Ak Mouse anti-Mouse, Brilliant Violet 421, Clone: 10-3.6, BD Optibuild™

Catalog No. BDB743471 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743471 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743471 Supplier BD Biosciences Supplier No. 743471

Mouse Monoclonal Antibody

The 10-3.6 monoclonal antibody specifically recognizes theβchain of the I-A[k] MHC class II alloantigen. It cross-reacts with splenocytes from mice of the H-2[f], H-2[r], and H-2[s] haplotypes, from nonobese diabetic (NOD, H-2[g7]) mice, and with lymph node cells of BN, DA, WRA, and WRC rats. Reactivity with other haplotypes ( e.g., b, d, p, q /) has not been observed. Long-term in vivo /treatment with 10-3.6 mAb has been reported to prevent the spontaneous development of autoimmune insulin-dependent diabetes mellitus (IDDM) in female NOD mice.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ak
Classification Monoclonal
Clone 10-3.6
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Aβk
Host Species Mouse
Immunogen C3H mouse splenocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.