Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ I-Ap Monoclonal Antibody (7-16.17)
GREENER_CHOICE

Catalog No. PIMA517767
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517767 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517767 Supplier Invitrogen™ Supplier No. MA517767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17767 targets I-Ap in CT and FACS applications and shows reactivity with Mouse samples. The MA5-17767 immunogen is b10.p.

MHC Class II (I-Ap) is a protein complex found on the surface of cells in vertebrates. Its main function is to present peptides derived from extracellular proteins to T cells for immune recognition. MHC Class II (I-Ap) is composed of an alpha and beta chain, both of which contribute to the peptide-binding groove. MHC Class II (I-Ap) is primarily restricted to antigen-presenting cells, including dendritic cells, macrophages, and B cells. These cells are responsible for capturing and processing antigens from the extracellular environment. Once the antigens are internalized, they are broken down into smaller peptides, which then bind to MHC Class II molecules within the endosomal compartments of these cells. MHC Class II (I-Ap) consists of an extracellular domain, a transmembrane domain, and a cytoplasmic tail. The extracellular domain is responsible for interacting with the antigenic peptides, while the transmembrane domain anchors the MHC Class II molecule to the cell membrane. The cytoplasmic tail plays a role in intracellular signaling events. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ap
Applications Cytotoxicity Assay, Flow Cytometry
Classification Monoclonal
Clone 7-16.17
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene H2-Eb1
Gene Accession No. P04230
Gene Alias cell surface glycoprotein; H-2 class II histocompatibility antigen, I-E beta chain; H2Eb; H-2Eb; H2-Eb1; histocompatibility 2, class II antigen E beta; Ia4; Ia-4; I-E beta MHC class II; major histocompatibility complex H-2E beta chain; MHC class II IE-b; MHC H2-IE-beta cell surface glycoprotein
Gene Symbols H2-Eb1
Host Species Mouse
Immunogen B10.p.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14969
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.