Learn More
Invitrogen™ BacLight™ Bacterial Membrane Potential Kit
Description
Specifications
Specifications
| Content And Storage | Contains DiOC2(3) (1.2 mL of 3 mM solution in DMSO), CCCP (300 μL of 500 μM solution in DMSO), and PBS (200 mL). Store in refrigerator (2–8°C) and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Membrane Potential Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Bacterial Membrane Potential Kit |
| Dye Type | Membrane Potential Probes |
| Format | Tube(s) |
| Product Line | BacLight |
| Quantity | 1 kit |
| Shipping Condition | Wet Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.