Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BacLight™ RedoxSensor™ CTC Vitality Kit

Catalog No. B34956
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34956 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34956 Supplier Invitrogen™ Supplier No. B34956
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The BacLight™ RedoxSensor™ CTC Vitality Kit provides effective reagents for evaluating bacterial cell health and vitality that can withstand fixation procedures.

The BacLight™ RedoxSensor™ CTC Vitality Kit provides effective reagents for evaluating bacterial cell health and vitality that can withstand fixation procedures. Briefly, healthy cells respiring via the electron transport chain will absorb and reduce CTC into an insoluble, red fluorescent formazan product. Cells not respiring or respiring at slower rates will reduce less CTC and consequently produce less fluorescent product, giving a semi-quantitative estimate of healthy vs. unhealthy bacteria. Green- and blue-fluorescent nucleic acid stains are included as counterstains to assist differentiation of cells from debris and calculating total cell numbers.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Bacteria
Content And Storage • 5-cyano-2,3-ditolyl tetrazolium chloride (CTC), 5 x 15 mg vials
• SYTO™ 24 green (100 μL in DMSO), 1 vial
• DAPI (5 mg/mL solution in water), 100 μL

Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type CTC Vitality Kit
Dye Type Other Label(s) or Dye(s)
Format Tube(s), Slide(s)
Product Line BacLight, RedoxSensor
Quantity 1 Kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Emission Wavelength 488
Show More Show Less
What bacterial parameters can I look at by flow cytometry?

You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.