Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ I-BET 151 hydrochloride
GSA_VAGREENER_CHOICESDP

Catalog No. 465050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-505-0 50 mg
46-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-505-0 Supplier R&D Systems™ Supplier No. 4650/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. BET bromodomain inhibitor; also promotes differentiation of hiPSCs into megakaryocytes

BET bromodomain inhibitor; blocks recruitment of BET to chromatin. Induces apoptosis and G0/G1 cell cycle arrest in MLL-fusion leukemic cell lines in vitro (IC50 values are 15, 26, 120 and 192 nM for NOMO1, MV4;11, MOLM13 and RS4;11 cell lines respectively); reduces BCL2 expression in NOMO1 cells. Improves survival in two rodent models of MLL-fusion leukemia in vivo. Enhances differentiation of human iPSC into megakaryocytes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

Molecular Formula C23H21N5O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 451.9
Purity >98%
Chemical Name or Material I-BET 151 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.