Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ I-BRD9
GREENER_CHOICE

Catalog No. 559110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
559110 10 mg
559150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 559110 Supplier Tocris Bioscience™ Supplier No. 5591/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective BRD9 inhibitor

I-BRD9 is a potent and selective BRD9 inhibitor (pIC50 = 7.3). Exhibits >70-fold selectivity for BRD9 over a panel of 34 other bromodomains and >700-fold selectivity over the BET family. Downregulates CLEC1, DUSP6, FES and SAMSN1 genes in Kasumi-1 cells.

Specifications

Chemical Name or Material 5-Ethyl-4,5-dihydro-4-oxo-N-(tetrahydro-1,1-dioxido-2H-thiopyran-4-yl)-7-[3-(trifluoromethyl)phenyl]thieno[3,2-c]pyridine-2-carboximidamide
CAS 1714146-59-4
Quantity 10 mg
Target Bromodomain Inhibitors
Molecular Formula C22H22F3N3O3S2
Formula Weight 497.55
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.