Learn More
Cytiva E. coli BL21
Catalog No. 45001328 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Vial
Catalog No. Quantity
45-001-328 1 Vial
1 options
Catalog No. 45-001-328 Supplier Cytiva Supplier No. 27154201
Add to Cart
Add to Cart
Protease-deficient E. coli host strain for optimal expression of recombinant proteins
  • F-, ompT, hsdS (rB-, mB-), gal, dcm
  • Exemplary host well suited for use with the GST gene fusion system
  • High-level expression of a variety of recombinant proteins
  • Defective in E. coli strain B, and wild-type restriction systems to permit high efficiency transformation with unmodified plasmid DNA
  • Verified by RAPD fingerprint analysis using Ready-To-Go™ RAPD analysis Kit
  • Lacks the major Lon protease responsible for degradation of damaged or foreign proteins
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Vial
Species E. coli