Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-E[κ] Mouse anti-Mouse, BV786, Clone: 17-3-3, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744680
Description
I-E[κ] Monoclonal antibody specifically detects I-E[κ] in Mouse samples. It is validated for Flow Cytometry.
Specifications
I-E[κ] | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
17-3-3 | |
Brilliant Violet 786 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction