Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli O157 Mouse anti-E. coli, Clone: 9.88, Abnova™

Catalog No. 89035106 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-106 Supplier Abnova Corporation Supplier No. MAB15086
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Escherichia coli O157.

  • High specificity
  • High reactivity

Specifications

Antigen Escherichia coli O157
Applications Immunofluorescence
Classification Monoclonal
Clone 9.88
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Escherichia coli O157.
Dilution Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Host Species Mouse
Immunogen A crude sonicate of Escherichia coli O157.
Purification Method PEG precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.