Learn More
Pyramex™ I-Force™ Safety Eyewear
Shop All Pyramex Safety Products Products
Click to view available options
Lens Tint:
Amber
Clear
Gray
Description
Specifications
Specifications
| Lens Type | H2X |
| Certifications/Compliance | ANSI Z87.1, CSA Z94.3-07 |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Gray |
| Frame Color | Black |
| Weight (Metric) | 48 g |
| Lens Material | Polycarbonate (Front Lens), Acetate (Inner Lens), PU Foam |
| Frame Material | Nylon |