Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giardia lamblia Mouse anti-Parasite, Clone: HM013, Abnova™

Catalog No. 89033513 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-513 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-513 Supplier Abnova Corporation Supplier No. MAB13550
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Giardia lamblia.

  • High specificity
  • High reactivity

Specifications

Antigen Giardia lamblia
Applications ELISA, Immunofluorescence, Lateral Flow, Western Blot
Classification Monoclonal
Clone HM013
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Giardia lamblia.
Dilution ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Host Species Mouse
Purification Method Protein A purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with antigen of Giardia lamblia.
Target Species Parasite
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.