Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, Biotin

Description
Includes
Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions
- Kits label any DNA or RNA template and are suitable for a wide range of applications
- One-step chemical method provides easy and consistent control of the labeling reactions
- Adjustable labeling density permits DNA and RNA to be optimally labeled for high sensitivity results
- Covalent mechanism provides permanent, nondestructive modification of nucleic acid residues
- Labels do not impact hybridization performance
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Biotin |
Final Product Type | DNA (Labeled), RNA (Labeled) |
Labeling Target | DNA (General), Oligos, RNA (General) |
For Use With (Application) | Efficient, direct, nonenzymatic labeling of DNA and RNA |
Includes | Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions |
Product Type | Label IT Biotin Labeling Kit |
Quantity | For 100 μg nucleic acid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.