Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Legionella pneumophila LPS (Serogroup 1), Mouse anti-L. pneumophila, Clone: AWB4CE4, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB18764
Specifications
Legionella pneumophila LPS (Serogroup 1) | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide). | |
LPS conjugated to BSA. | |
RUO | |
L. pneumophila | |
Antibody | |
IgG1 κ |
ELISA | |
AWB4CE4 | |
ELISA The optimal working dilution should be determined by the end user. | |
Mouse | |
20 μg | |
Primary | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction