Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella pneumophila LPS (Serogroup 1), Mouse anti-L. pneumophila, Clone: AWB4CE4, Abnova™

Catalog No. 89335597 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-597 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-597 Supplier Abnova Corporation Supplier No. MAB18763
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen Legionella pneumophila LPS (Serogroup 1)
Applications ELISA
Classification Monoclonal
Clone AWB4CE4
Conjugate Unconjugated
Dilution ELISA The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide).
Host Species Mouse
Immunogen LPS conjugated to BSA.
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species L. pneumophila
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.