Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mirVana™ miRNA Mimic, Negative Control #1

mirVana miRNA Mimic Negative Control No. 1 is a non-targeting control for experiments with mirVana miRNA mimics. Providing a baseline for evaluating miRNA gene expression effects, this miRNA mimic negative control is validated to ensure no impact on known miRNA functions in human cells.

$333.06 - $2802.80

Specifications

Content And Storage Store at or below –20°C. Do not store in a frost-free freezer.
Control Type Negative Control
RNAi Type miRNA Mimic
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
44-640-58
Encompass_Preferred
View Documents
Invitrogen™
4464058
5 nmol
Each for $333.06

Save 9%

Reg : $366.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
 
44-640-59
Encompass_Preferred
View Documents
Invitrogen™
4464059
20 nmol
Each for $960.96

Save 9%

Reg : $1,056.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
44-640-60
Encompass_Preferred
View Documents
Invitrogen™
4464060
50 nmol
Each for $1,550.00
Only null left
Add to Cart
 
44-640-61
Encompass_Preferred
View Documents
Invitrogen™
4464061
250 nmol
Each for $2,802.80

Save 9%

Reg : $3,080.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
Description

Description

mirVana miRNA Mimic Negative Control No. 1 is a non-targeting control for experiments with mirVana miRNA mimics. Transfected using the same methodology as positive controls and experimental miRNA mimics, it provides a baseline for evaluating effects on target gene expression. Extensively tested in human cell lines and tissues, it ensures no impact on known miRNA functions, making it essential for accurate gene expression studies. Key properties include reliability, superior specificity, and versatile use in vitro and in vivo.

Features of mirVana miRNA Mimic Negative Control include:

  • Reliability—maximum and consistent effect in vitro at low concentration and low toxicity
  • Superior specificity—the most updated Sanger content with unique Star strand modification
  • Versatile use—can be used in vitro and in vivo, offering you consistency throughout your entire research project

What are mirVana miRNA mimics?

mirVana miRNA mimics are small, chemically modified double-stranded RNA molecules designed to mimic endogenous miRNA molecules and enable miRNA functional analysis by up-regulation of miRNA activity. Positive and negative controls are also available for use along with mirVana miRNA mimics in miRNA up-regulation experiments.

Quality control

Each mirVana miRNA Mimic is synthesized and purified in state-of-the-art facilities to meet the highest quality standards. Our rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. We also assess each annealed miRNA mimic by gel electrophoresis to ensure proper strand annealing. The result is a premium-quality miRNA mimic that is purified and ready for use.

Specifications

Specifications

Store at or below –20°C. Do not store in a frost-free freezer.
miRNA Mimic
Unconjugated
miRNA Mimic
Negative Control
miRNA & Non-Coding RNA Analysis
mirVana™
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.