Learn More
Thermo Scientific™ I-PER™ Insect Cell Protein Extraction Reagent PROMO
Description
The baculovirus insect cell expression system is an efficient and popular system for the production of recombinant (eukaryotic) proteins in cell culture. Proteins expressed in baculoviral systems can be used for structural analyses, biochemical assays and a variety of other applications. I-PER Reagent maintains functionality of extracted proteins and is directly compatible with downstream applications such as protein assays, Western blotting and 6xHis-tagged protein purification.
Highlights:
Gentle extraction – optimized, mild non-ionic detergent provides maximum extraction of soluble proteins from insect cells
Effective – provides better protein extraction than sonication
Compatible – downstream compatibility with Western blotting, 6xHis-tagged protein purification, protein assays and ion-exchange chromatography
Flexible – useful for protein extraction from suspended or adherent cultured insect cells
Specifications
Specifications
Quantity | 250 mL |
Format | Liquid |
Product Type | Reagent |
Volume (Metric) | 250 mL |
Content And Storage | Upon receipt store at 4°C. |
Product Line | I-PER™ |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.