Learn More
Applied Biosystems™ Power SYBR™ Green RNA-to-CT™ 1-Step Kit
Description
The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.
Get sensitivity and specificity in an easy-to-use one-step qRT-PCR reaction. The Power SYBR™ Green RNA-to-CT™ 1-Step kit has been formulated to ensure maximum sensitivity and reliability. The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets. It includes:
- AmpliTaq™ Gold DNA Polymerase UP for hot start qPCR, minimizing non-specific product formation
- ArrayScript™ UP Reverse Transcriptase, an engineered RT that produces high yields of full-length cDNA
- An additive that reduces primer dimer formation, reducing non-specific products that can cause false positive signals
- RNase inhibitor, to prevent degradation of RNA templates
- A proprietary ROX™ dye that serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations for superb precision on Applied Biosystems™ real-time PCR instruments.
Order Info
Shipping Conditions: Wet Ice
Specifications
Specifications
| Content And Storage | Sufficient for 500 reactions at 20 μL or 200 reactions at 50 μL reaction volume and includes: • 1 x 5 mL tube of 2X master mix containing SYBR™Green 1 Dye, AmpliTaq Gold™ DNA Polymerase UP, dNTPs, Passive Reference 1,and optimized buffer components. Store at 2-8°C after opening. • 1 x 80 μL tube of 125X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase, RNase Inhibitor. Store at -15 to -30°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Detection Method | SYBR |
| PCR Method | 1-step RT-qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System |
| Label or Dye | SYBR Green |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | RNA-to-CT, SYBR |
| Product Type | One-Step qRT-PCR Kit |
| Show More |
Frequently Asked Questions (FAQs)
If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:
- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)
Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).
In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).
There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.
There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.