Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ i-Shields™ Eyeshields
SDP

Catalog No. 22500451 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Assorted
Clear
No. per Pack:
10/Pk.
100/Cs.
100/Pk.
25/Pk.
LABPAK Professional Pack (25 Lenses and 10 Frames)
Product Type:
Eyeshield frame
Eyeshield frame and lens
Eyeshield frames
Eyeshield lens
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack Product Type
22-500-451 Clear 100/Pk. Eyeshield lens
22-500-453 Assorted 10/Pk. Eyeshield frame
22-500-450 Clear 25/Pk. Eyeshield lens
22-500-454 Assorted 100/Cs. Eyeshield frames
22-500-452 Assorted LABPAK Professional Pack (25 Lenses and 10 Frames) Eyeshield frame and lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-500-451 Supplier Cancer Diagnostics, Inc.™ Supplier No. ES0100
Add to Cart
Add to Cart

Includes

Eyeshield LENS only. (Frame is sold separately.)

Can be worn with a surgical mask.

  • Ultra Lightweight for stress free/mark free extended wearing
  • Reusable frame and disposable lenses provide side shield protection
  • Distortion-free, optical grade lenses work well with most sizes of prescription eyewear and can be worn with a surgical mask
  • Frames packaged in a variety of colors
  • Lightweight recyclable components can be easily sterilized

Order Info

NOTE: Lenses and frames are ordered separately unless ordered in combo pack (such as 22-500-452).

Specifications

Color Clear
Product Type Eyeshield lens
For Use With (Equipment) most sizes of prescription eyewear and can be worn with a surgical mask
No. per Pack 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.