Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Silencer™ GAPDH siRNA (human, mouse, rat)

Silencer™ GAPDH siRNA (human, mouse, rat)

$224.46 - $1296.00

Specifications

Control Type Positive Control
RNAi Type siRNA
Species Human, Mouse, Rat
Label or Dye Unconjugated
Product Line Silencer™, Ambion™
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AM4624
Encompass_Preferred
View Documents
Invitrogen™
AM4624
5 nmol
Each for $224.46

Save 14%

Reg : $261.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
AM4631
Encompass_Preferred
View Documents
Invitrogen™
AM4631
40 nmol
Each for $574.74

Save 7%

Reg : $618.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart
 
AM4632
Encompass_Preferred
View Documents
Invitrogen™
AM4632
5 x 40 nmol
Each for $1,296.00
Only null left
Add to Cart
 
Description

Description

Ambion™ Silencer™ GAPDH Positive Control siRNA (human, mouse, rat) is ideal for developing and optimizing siRNA experiments. The control is validated for use in human, mouse, and rat cell lines.
  • Validated siRNA controls for optimizing siRNA experiments
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs.
If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
Specifications

Specifications

Positive Control
Human, Mouse, Rat
Silencer™, Ambion™
HPLC
siRNA
Unconjugated
siRNA
GAPDH
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.