Learn More
Invitrogen™ Silencer™ Select Negative Control No. 1 siRNA
Silencer Select Negative Control No. 1 is a bioinformatically designed, non-targeting siRNA crucial for siRNA experiments. Matching Silencer Select chemistry, it ensures minimal off-target effects and is an essential siRNA negative control for achieving clear, accurate gene silencing results
$256.62 - $674.24
Specifications
Control Type | Negative Control |
---|---|
RNAi Type | siRNA |
For Use With (Application) | RNAi, Transfection |
Label or Dye | Unconjugated |
Product Line | Silencer™, Silencer™ Select, Ambion™ |
Description
Silencer Select Negative Control No. 1 siRNA is a bioinformatically designed, non-targeting siRNA used as a negative control siRNA. It distinguishes specific effects of siRNA-mediated gene silencing from non-specific procedural effects, ensuring minimal off-target effects and providing a reliable baseline for comparison. Validated through extensive testing, it has minimal impact on gene expression, cell proliferation, viability, and morphology, making it essential for accurate results in human, mouse, and rat cells.
Negative controls, such as non-targeting siRNAs, are critical for distinguishing the specific effects of siRNA-mediated gene silencing from non-specific effects caused by the experimental procedure itself. Silencer Select negative control No. 1 is designed to ensure minimal off-target effects, providing a robust baseline for comparing the results of siRNA-treated samples. This negative control siRNA shares the same chemical modifications as other Silencer Select siRNAs, ensuring enhanced efficacy and specificity.
Properties of Silencer Select Negative Control siRNAs include:
- No significant sequence similarity to human mouse, or rat genes
- Minimal effects on cell proliferation and viability
- Silencer Select modifications for enhanced specificity
Validated through extensive testing, including microarray analysis and multi-parametric cell-based assays, Silencer Select Negative Control No. 1 has been proven to have minimal impact on gene expression, cell proliferation, viability, and morphology. This makes it an essential tool for researchers aiming to achieve accurate and consistent phenotypic data in human, mouse, and rat cells.
What are Silencer Select siRNAs?
Silencer Select siRNAs represent our next-generation siRNAs, incorporating the latest advancements in siRNA design, off-target effect prediction algorithms, and chemical modifications. These improvements result in siRNAs with unrivalled efficacy, potency, and specificity, leading to fewer failed experiments and more consistent, cleaner phenotypic data.
Quality control
Each Silencer Select siRNA is synthesized and purified in state-of-the-art facilities to meet the highest quality standards. Our rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. We also assess each annealed siRNA by gel electrophoresis to ensure proper strand annealing. The result is a premium-quality siRNA that is purified and ready for use.
Specifications
Negative Control | |
RNAi, Transfection | |
Silencer™, Silencer™ Select, Ambion™ | |
HPLC |
siRNA | |
Unconjugated | |
siRNA | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.