Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Silencer™ siRNA Construction Kit with manual

Catalog No. p-7180302
Encompass_Preferred
Click to view available options
Includes:
Kit only
Kit with manual

Catalog No. AM1620

Supplier: Invitrogen™ AM1620

Only null left
Add to Cart
Add to Cart
The Invitrogen™ Silencer™ siRNA Construction Kit is for the synthesis of siRNAs by in vitro transcription of DNA templates and costs less per siRNA than chemical synthesis. The kit includes sufficient reagents to synthesize 15 siRNAs.
  • Cut siRNA preparation time—generate up to 15 siRNAs in less than 24 hours
  • Get hundreds of transfections per reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 2X
    Content And Storage DNA Hybridization Buffer, T7 Promoter Primer, Klenow Reaction Buffer, 10X dNTP, Exo– Klenow, T7 Enzyme Mix, T7 Reaction Buffer, 2X NTP Mix, Digestion Buffer, DNase, RNase, Sense Control DNA, and Antisense Control DNA should be stored at –20°C. siRNA Binding Buffer, siRNA Wash Buffer, Filter Cartridges, and Tubes should be stored at room temperature. Nuclease-free Water may be stored at any temperature.
    Control Type Transcription Control
    Sample Type In vitro Transcribed siRNA
    Includes Kit only
    No. of Reactions 15
    Product Line Silencer, Ambion
    Quantity 15 reactions
    Shipping Condition Dry Ice
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.