Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus aureus Rabbit anti-Staphylococcus, Polyclonal, Abnova™

Catalog No. 89033658 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-658 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-658 Supplier Abnova Corporation Supplier No. PAB29813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against Staphylococcus aureus.

  • High specificity
  • High reactivity

Specifications

Antigen Staphylococcus aureus
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Staphylococcus aureus.
Dilution ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Formulation In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Host Species Rabbit
Immunogen Staphylococcus aureus
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with all antigens of Staphylococcus aureus. May cross-react with related microbes.
Target Species Staphylococcus species
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.