Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Alcohol, ACS, 99.0%, MilliporeSigma™

Catalog No. 50947978 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging:
Plastic Bottle
Tinplate Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-947-978 Tinplate Metal Can
50-948-022 Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-947-978 Supplier MilliporeSigma Supplier No. BX18051
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 24 to 25°C
Quantity 1 L
Flash Point 14°C
Packaging Tinplate Metal Can
Residue after Evaporation 0.003% max
Assay Percent Range 99.0% min.
Vapor Pressure 40.7 hPa (20°C)
Boiling Point 81°C to 83°C (1013 hPa)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.