Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Toxoplasma gondii IgA ELISA Kit

Catalog No. 89032310 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit

Enzyme-linked immunosorbent assay for the detection of IgA antibodies to Toxoplasma gondii.

Intended for detection of specific IgA antibodies in a sample by means of a sandwich type of the EIA method (i.e. a solid phase coated with specific antigen – antibody from the analyzed sample – labeled antibody).

Specifications

Description Toxoplasma gondii IgA ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgA antibodies to Toxoplasma gondii.
Conjugate HRP
Detection Method Colorimetric
Product Type Toxoplasma gondii IgA ELISA Kit
Sample Type Human Plasma, Human Serum
Target Species Human
For Use With (Application) Qualitative
Kit Contents Microtitre Plate: 96 wells, Negative Control: (2mL), CUT-OFF solution: (3mL), Positive Control: (2mL), Tracer: Lyophilisate containing T. gondii antigen and conjugate, Conjugate: 15mL, Sample Diluent 3 (105mL), TMB-Complete 2: 15mL, Tracer Diluent (13mL), Wash Solution: 20x concentrated buffer: 75mL, Stop Solution: Acid solution: 15mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at 4°C. Do not freeze.
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.