Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-HeLaMONSTER™ Transfection Kit

Catalog No. MIR2904 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 x 0.4 mL
1 x 1 mL
5 x 1 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2904 1 x 0.4 mL
MIR2905 5 x 1 mL
MIR2906 10 x 1 mL
MIR2900 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2904 Supplier Mirus Bio™ Supplier No. MIR2904
Add to Cart
Add to Cart

High-efficiency, low-toxicity DNA transfection kit optimized for HeLa cells and associated cell lineages. This kit is suitable for both transient and stable transfection. Transfection does not require medium changes and can be performed in the presence of serum.

  • Each kit contains a base reagent and a booster reagent to optimize transfection performance in HeLa cells
  • Low cellular toxicity maintains cell density and reduces experimental bias
  • Allows up to 50-60% transfection efficiency in HeLa cells
  • Delivers single or multiple plasmids for applications including gene expression and promoter analysis
  • Concentration: 1.33 mg/mL

Specifications

Content And Storage Store at -20°C
Cell Type Tumor Cell Lines
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 1.33 mg/mL
For Use With (Application) Gene expression and promoter analysis, optimized to provide superior transfection efficiency in HEK 293 cells without sacrificing cellular health
Includes Base and booster reagent
Product Type Transfection Reagent
Quantity 1 x 0.4 mL
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.