Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-PRO™ Transfection Reagent

Catalog No. p-6629085
Click to view available options
Quantity:
1 mL
10 mL

TransIT-PRO™ Transfection Reagent decreases time to produce usable protein by maximizing target protein yields through transient transfection; high, reproducible protein yield in suspension CHO and 293 cells.

Reach critical stages faster. Decrease time to produce usable protein by maximizing target protein yields through transient transfection. The TransIT-PRO™ Transfection Reagent uses animal-origin-free components designed for high and reproducible protein yield in suspension CHO and 293 derived cells. Since it is compatible with varied media formulations, the same media can be used for both transient and stable expression. TransIT-PRO™ outperforms linear PEI in protein yield, while providing a cost-effective alternative to FreeStyle™ MAX.
  • Suitable for both transient and stable transfection
  • Compatible with multiple CHO media formulations
  • Reproducible protein expression with minimal optimization
  • Outperforms linear PEI in protein yield and provides a cost-effective alternative to FreeStyle™ MAX

Warranty and Services

1 Year

Specifications

Content And Storage Store at -20°C
Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
Product Type Transfection Reagent
Quantity 1 mL
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.