Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IA-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540471 Supplier Invitrogen™ Supplier No. PA540471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PAAALRPPGK RPPPPTAAEP PAKAVKAPGA KKPKAIRKLH FEDEVTTSPV Sequence homology: Cow: 100%; Dog: 100%; Horse: 79%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 79%; Zebrafish: 93%.

IA-1, also known as INSM1, is an essential five zinc-finger transcription factor that while initially identified from an insulinoma cDNA library, is expressed in the developing nervous system. Specifically, IA-1 is expressed as early as E9.5 in mice in the fore-, mid- and hindbrain, spinal cord, retina, and olfactory bulb. It is thought that IA-1 functions as a transcriptional repressor, and is a part of several signaling pathways including those of Notch and sonic hedgehog in addition to that of Ngn3 during pancreatic endocrine cell differentiation. IA-1 is also expressed in multiple tumors, including the majority of neuroendocrine tumors tested so far as well as nearly 100% of all small cell lung carcinomas, indicating that IA-1 may be an important target in cancer therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IA-1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene INSM1
Gene Accession No. Q01101
Gene Alias IA1; IA-1; INSM transcriptional repressor 1; Insm1; insulinoma associated 1; insulinoma-associated 1; insulinoma-associated protein (IA-1); insulinoma-associated protein 1; insulinoma-associated protein 1-like; LOC100912563; Zinc finger protein IA-1
Gene Symbols INSM1
Host Species Rabbit
Immunogen Immunogen peptide is a 14 amino acid sequence within the following: PAAALRPPGK RPPPPTAAEP PAKAVKAPGA KKPKAIRKLH FEDEVTTSPV (aa 191-240).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3642
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.