Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IA-2/PTPRN Mouse anti-Human, Clone: 1D3F5, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 660451IG150UL

Description
IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.Specifications
IA-2/PTPRN | |
Monoclonal | |
1.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16849 | |
PTPRN | |
IA-2/PTPRN Fusion Protein Ag17909 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG3 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1D3F5 | |
Unconjugated | |
PTPRN | |
IA 2/PTP, IA2, IA-2, ICA 512, ICA3, ICA512, Islet cell antigen 512, Islet cell autoantigen 3, PTP IA 2, PTPRN, R PTP N | |
Mouse | |
Protein A | |
RUO | |
5798 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction