Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IA-6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560857 Supplier Invitrogen™ Supplier No. PA560857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHLSTHEAGS ARALAPGFGS ERGAPLAFAC PLCGAHFPTA DIREKHRLWH AVREELLLPA LAGAPPETSG PSGPSDGSAQ QIFSCKHCPS TF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

IA-6, also known as insulinoma-associated 2 or INSM2, is related to IA-1 (INSM1), a zinc-finger factor that is expressed in the developing nervous system. While the exact function of IA-6 is not known, other related insulinoma-associated proteins such as IA-2 are major autoantigens in type 1 diabetes. The homology exhibited by IA-6 to IA-1 has led to speculation that IA-6 may also play a role in the regulation of gene transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IA-6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INSM2
Gene Accession No. Q96T92
Gene Alias IA6; IA-6; INSM transcriptional repressor 2; Insm2; INSM2 variant 1; insulinoma-associated 2; insulinoma-associated protein 2; insulinoma-associated protein IA-6; methylated in liver tumor 1; mlt 1; mlt1; Nbla106; Zinc finger protein IA-6
Gene Symbols INSM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IA-6. Recombinant protein control fragment (Product #RP-100071).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.