Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552396 Supplier Invitrogen™ Supplier No. PA552396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSYGDLPGPS PAQLFQDSGL LYLAQELPAP SRARVPRLPE QGSSSRAEDS PEGYEKEGLG DRGEKPASPA VQPDAALQRL AAVLAGYGVE LRQLTPEQLS TLLTLLQLLP KGA Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 81%.

IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPRN
Gene Accession No. Q16849
Gene Alias 105 kDa islet cell antigen; BEM-3; brain-enriched membrane-associated protein tyrosine phosphatase; IA2; IA-2; IA-2/PTP; ICA 512; ICA105; ICA3; ICA512; ICA512-CCF; ICA512-cleaved cytosolic fragment; ICA512-N-terminal fragment; ICA512-NTF; ICA512-TMF; ICA512-transmembrane fragment; islet cell antigen 2; islet cell antigen 512; Islet cell autoantigen 3; mIA-A; mKIAA4064; protein tyrosine phosphatase, receptor type N; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase, receptor-type, N; protein tyrosine phosphatase-like N; PTP IA-2; PTPLP; Ptprn; receptor-type tyrosine-protein phosphatase-like N; R-PTP-N
Gene Symbols PTPRN
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PTPRN (Product #RP-88661)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5798
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.