Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592949 Supplier Invitrogen™ Supplier No. PA592949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STGHMILAYM EDHLRNRDRL AKEWQALCAY QAEPNTCATA QGEGNIKKNR HPDFLPYDHA RIKLKVESSP SRSDYINASP IIEHDPRMPA YIATQGPLSH TIADFWQMVW ESGCTVIVML TPLVEDGVKQ CDRYWPDEGA SLYHVYEVNL VSEHIWCEDF LVRSFYLKNV QTQETRTLTQ FHFLSWPAEG TPASTRPLLD FRRKVNKCYR GRSCPIIVHC SDGAGRTGTY ILIDMVLNRM AKGVKEIDIA ATLEHVRDQR PGLVRSKDQF EFALTAVAEE VNAILKALPQ; Positive Samples: 293T, NIH/3T3, Raji, Mouse brain, Rat brian; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasmic vesicle, Endosome, Membrane, Perikaryon, Single-pass type I membrane protein, axon, secretory vesicle membrane, synapse.

IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTPRN
Gene Accession No. Q16849, Q60673, Q63259
Gene Alias 105 kDa islet cell antigen; BEM-3; brain-enriched membrane-associated protein tyrosine phosphatase; IA2; IA-2; IA-2/PTP; ICA 512; ICA105; ICA3; ICA512; ICA512-CCF; ICA512-cleaved cytosolic fragment; ICA512-N-terminal fragment; ICA512-NTF; ICA512-TMF; ICA512-transmembrane fragment; islet cell antigen 2; islet cell antigen 512; Islet cell autoantigen 3; mIA-A; mKIAA4064; protein tyrosine phosphatase, receptor type N; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase, receptor-type, N; protein tyrosine phosphatase-like N; PTP IA-2; PTPLP; Ptprn; receptor-type tyrosine-protein phosphatase-like N; R-PTP-N
Gene Symbols PTPRN
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116660, 19275, 5798
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.