Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IAA6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144534
Description
IAA6 Polyclonal Antibody for Western Blot, ELISA
Aux/IAA proteins are short-lived transcriptional factors that function as repressors of early auxin response genes at low auxin concentrations. Repression is thought to result from the interaction with auxin response factors (ARFs), proteins that bind to the auxin-responsive promoter element (AuxRE). Formation of heterodimers with ARF proteins may alter their ability to modulate early auxin response genes expression.
Specifications
IAA6 | |
Polyclonal | |
Unconjugated | |
IAA6 | |
SHY1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
841717 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.21 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q38824 | |
IAA6 | |
Recombinant Arabidopsis thaliana IAA6 protein (46-91aa). | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction