Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IAH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565270 Supplier Invitrogen™ Supplier No. PA565270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRLLLFGDSIT QFSFQQGGWG ASLADRLVRK CDVLNRGFSG YNTRWAKIIL PRLIRKGNSL DIPVAVTIFF GANDSALKDE NPK Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IAH1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Iah1
Gene Accession No. Q2TAA2
Gene Alias 1500019E20Rik; 4833421E05Rik; Harpb64; hypertrophic agonist responsive protein B64; hypertrophic agonist-responsive protein B64; Iah1; Isoamyl acetate-hydrolyzing esterase 1 homolog; isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae); testicular tissue protein Li 92
Gene Symbols Iah1
Host Species Rabbit
Immunogen Recombinant Human IAH1. Recombinant protein control fragment (Product #RP-96079).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.