Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
iap, Mouse anti-Listeria sp., Clone: p6017, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB15881
Specifications
iap | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4. | |
- | |
Mouse | |
100 μg | |
Primary | |
L. monocytogenes | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
p6017 | |
ELISA (1:5000-1:10000) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user. | |
P60 extracellular protein, invasion associated protein Iap | |
iap | |
Recombinant protein corresponding to Listeria monocytogenes iap. | |
RUO | |
985140 | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction