Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544246 Supplier Invitrogen™ Supplier No. PA544246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SKHKPKFTFY DGPPFATGLP HYGHILAGTI KDIVTRYAHQ SGFHVDRRFG Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 93%; Rat: 86%; Yeast: 100%; Zebrafish: 79%.

IARS is a member of aminoacyl-tRNA synthetases which catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Isoleucine-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family and has been identified as a target of autoantibodies in the autoimmune disease polymyositis/dermatomyositis. Two alternatively spliced variants have been isolated that represent alternate 5' UTRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IARS
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IARS1
Gene Accession No. P41252
Gene Alias 2510016L12Rik; AI327140; AU044614; E430001P04Rik; fi46h05; FLJ20736; IARS; IARS1; Iarsl; ILERS; ILRS; IRS; isoleucine tRNA ligase 1, cytoplasmic; isoleucine tRNA synthetase; isoleucine--tRNA ligase, cytoplasmic; isoleucine-tRNA synthetase; isoleucyl tRNA synthetase; isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase 1; isoleucyl-tRNA synthetase, cytoplasmic; PRO0785; wu:fi46h05; zgc:63790
Gene Symbols IARS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human IARS (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3376
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.