Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IARS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578012
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (92%), Xenopus laevis (80%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IARS is a member of aminoacyl-tRNA synthetases which catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Isoleucine-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family and has been identified as a target of autoantibodies in the autoimmune disease polymyositis/dermatomyositis. Two alternatively spliced variants have been isolated that represent alternate 5' UTRs.
Specifications
IARS | |
Polyclonal | |
Unconjugated | |
IARS1 | |
2510016L12Rik; AI327140; AU044614; E430001P04Rik; fi46h05; FLJ20736; IARS; IARS1; Iarsl; ILERS; ILRS; IRS; isoleucine tRNA ligase 1, cytoplasmic; isoleucine tRNA synthetase; isoleucine--tRNA ligase, cytoplasmic; isoleucine-tRNA synthetase; isoleucyl tRNA synthetase; isoleucyl-tRNA synthetase; isoleucyl-tRNA synthetase 1; isoleucyl-tRNA synthetase, cytoplasmic; PRO0785; wu:fi46h05; zgc:63790 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
306804, 3376 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P41252 | |
IARS1 | |
Recombinant protein encompassing a sequence within the center region of human IARS. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction